Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555475 Supplier Thermo Scientific Supplier No. PA555475
Add to Cart
Add to Cart

PARP6 Polyclonal Antibody

PARP6 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Parp6
Gene Accession No. Q2NL67
Gene Alias 1700119G14Rik; 2310028P13Rik; 3110038K10Rik; ADP-ribosyltransferase diphtheria toxin-like 17; ARTD17; C030013N01Rik; PARP6; PARP-6; PARP-6-B1; PARP-6-C; pART17; poly (ADP-ribose) polymerase family, member 6; poly [ADP-ribose] polymerase 6; poly(ADP-ribose) polymerase family member 6; Protein mono-ADP-ribosyltransferase PARP6; RGD1310937
Gene Symbols Parp6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PARP6. Recombinant protein control fragment (Product #RP-90670).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.