Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155363 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155363 Supplier R&D Systems Supplier No. NBP155363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARP7 Polyclonal specifically detects PARP7 in Human samples. It is validated for Western Blot.

Specifications

Antigen PARP7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q7Z3E1
Gene Alias DDF1, DKFZP434J214, DKFZp686N0351, DKFZp686P1838, EC 2.4.2.30, FLJ40466, PARP-1, PARP-7, PARP7DKFZp434J214, pART14, Poly [ADP-ribose] polymerase 7, RM1, TCDD-inducible poly [ADP-ribose] polymerase, TCDD-inducible poly(ADP-ribose) polymerase
Gene Symbols TIPARP
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TIPARP(TCDD-inducible poly(ADP-ribose) polymerase) The peptide sequence was selected from the N terminal of TIPARP. Peptide sequence LKTCFKKKDQKRLGTGTLRSLRPILNTLLESGSLDGVFRSRNQSTDENSL.
Molecular Weight of Antigen 76 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 25976
Test Specificity Mouse: 83%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.