Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550719 Supplier Invitrogen™ Supplier No. PA550719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PARP8 protein.

Poly(ADP-ribosylation) is a method of DNA damage-dependent posttranslational modification that helps to rescue injured proliferating cells from cell death. The PARP (poly(ADP-ribose) polymerase) proteins comprise a superfamily of enzymes that functionally modify histones and other nuclear proteins, thereby preventing cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PARP8
Gene Accession No. Q3UD82, Q8N3A8
Gene Alias 2810430O08Rik; ADP-ribosyltransferase diphtheria toxin-like 16; ARTD16; D13Ertd275e; I79_004209; Parp8; PARP-8; pART16; poly (ADP-ribose) polymerase family, member 8; poly [ADP-ribose] polymerase 8; poly(ADP-ribose) polymerase family member 8; Protein mono-ADP-ribosyltransferase PARP8
Gene Symbols PARP8
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human poly (ADP-ribose) polymerase family, member 8.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52552, 79668
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.