Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113582 Supplier Invitrogen™ Supplier No. PA5113582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHGGGLALAL VKAGGFEIQE ESKQFVARYG KVSAGEIAVT GAGRLPCKQI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

In concert with DTX3L plays a role in PARP1-dependent DNA damage repair. PARP1-dependent PARP9/BAL1-DTX3L-mediated ubiquitination promotes the rapid and specific recruitment of 53BP1/TP53BP1, UIMC1/RAP80, and BRCA1 to DNA damage sites. Involved in inducing the expression of IFN-gamma-responsive genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PARP9
Gene Accession No. Q8IXQ6
Gene Alias ADP-ribosyltransferase diphtheria toxin-like 9; ARTD9; AW214463; b aggressive lymphoma protein; B aggressive lymphoma protein homolog; Bagl; Bal; BAL1; BC003281; MGC:7868; Parp9; PARP-9; poly (ADP-ribose) polymerase 9; poly (ADP-ribose) polymerase family, member 9; poly [ADP-ribose] polymerase 9; poly(ADP-ribose) polymerase family member 9; Protein mono-ADP-ribosyltransferase PARP9; RGD1307534
Gene Symbols PARP9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human PARP9 (aa 144-193).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83666
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.