Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP9 Rabbit anti-Human, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501731631 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1631 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1631 Supplier Proteintech Group Inc Supplier No. 175351AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In concert with DTX3L plays a role in PARP1-dependent DNA damage repair. PARP1-dependent PARP9/BAL1-DTX3L-mediated ubiquitination promotes the rapid and specific recruitment of 53BP1/TP53BP1, UIMC1/RAP80, and BRCA1 to DNA damage sites. Involved in inducing the expression of IFN-gamma-responsive genes.

Specifications

Antigen PARP9
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PARP9
Gene Accession No. Q8IXQ6
Gene Alias B aggressive lymphoma protein, BAL, BAL1, PARP 9, PARP9, Poly [ADP ribose] polymerase 9
Gene Symbols PARP9
Host Species Rabbit
Immunogen PARP9 Fusion Protein Ag11587
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303905, 83666
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.