Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARS2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219702
Description
PARS2 Polyclonal specifically detects PARS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PARS2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
DKFZp727A071, EC 6.1.1.15, MGC14416, MGC19467, MT-PRORS, probable prolyl-tRNA synthetase, mitochondrial, proline tRNA ligase 2, mitochondrial (putative), Proline--tRNA ligase, prolyl-tRNA synthetase (mitochondrial)(putative), prolyl-tRNA synthetase 2, mitochondrial (putative), prolyl-tRNA synthetase, mitochondrial (putative), proRS | |
Rabbit | |
53 kDa | |
0.1 mL | |
Lipid and Metabolism | |
25973 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q7L3T8 | |
PARS2 | |
Recombinant protein encompassing a sequence within the C-terminus region of human PARS2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Species reactivity based on sequence identity: Mouse (84%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction