Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARS2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PARS2 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PARS2 Polyclonal specifically detects PARS2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PARS2 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
25973 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RFHHCAPRRGRRLLLSRVFQPQNLREDRVLSLQDKSDDLTCKSQRLMLQVGLIYPASPGCYHLLPYTVRAMEKLVRVI | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
DKFZp727A071, EC 6.1.1.15, MGC14416, MGC19467, MT-PRORS, probable prolyl-tRNA synthetase, mitochondrial, proline tRNA ligase 2, mitochondrial (putative), Proline--tRNA ligase, prolyl-tRNA synthetase (mitochondrial)(putative), prolyl-tRNA synthetase 2, mitochondrial (putative), prolyl-tRNA synthetase, mitochondrial (putative), proRS | |
PARS2 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title