Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595781 Supplier Invitrogen™ Supplier No. PA595781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HHCAPRRGRR LLLSRVFQPQ NLREDRVLSL QDKSDDLTCK SQRLMLQVGL IYPASPGCYH LLPYTVRAME KLVRVIDQEM QAIGGQKVNM PSLSPAELWQ ATNRWDLMGK ELLRLRDRHG KEYCLGPTHE EAITALIASQ KKLSYKQLPF LLYQVTRKFR DEPRPRFGLL RGREFYMKDM YTFDSSPEAA Q; Positive Samples: Rat brain; Cellular Location: Mitochondrion matrix.

This gene encodes a putative member of the class II family of aminoacyl-tRNA synthetases. These enzymes play a critical role in protein biosynthesis by charging tRNAs with their cognate amino acids. This protein is encoded by the nuclear genome but is likely to be imported to the mitochondrion where it is thought to catalyze the ligation of proline to tRNA molecules. Mutations have been found in this gene in some patients with Alpers syndrome. [provided by RefSeq, Mar 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PARS2
Gene Accession No. Q5M7W7
Gene Alias BC027073; class II tRNA synthetase; MT-PRORS; PARS2; Probable proline--tRNA ligase, mitochondrial; probable prolyl-tRNA synthetase, mitochondrial; proline tRNA ligase 2, mitochondrial (putative); proline--tRNA ligase; Prolyl-tRNA synthetase; prolyl-tRNA synthetase (mitochondrial)(putative); prolyl-tRNA synthetase 2, mitochondrial (putative); proRS; RGD1305345
Gene Symbols PARS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-220 of human PARS2 (NP_6894812).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313429
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.