Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parter Medical Products™ Sterile Specimen Containers

Catalog No. 22150326 Shop All Parter Medical Products Products
Change view
Click to view available options
Capacity (Metric):
90 mL
Capacity (English):
3.0 oz.
Closure Color:
Blue
Natural
Orange
No. per Case:
100/Cs.
400/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Capacity (English) Closure Color No. per Case
22-150-326 90 mL 3.0 oz. Natural 400/Cs.
22-150-327 90 mL 3.0 oz. Natural 100/Cs.
22-150-328 90 mL 3.0 oz. Blue 400/Cs.
22-150-330 90 mL 3.0 oz. Blue 100/Cs.
22-150-329 90 mL 3.0 oz. Orange 400/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-150-326 Supplier Parter Medical Products™ Supplier No. 243016
Add to Cart
Add to Cart
This item is not returnable. View return policy

Incorporate the proprietary PMP™ Sure-Seal™ integrated sealing system and the proprietary PMP Sure-Lok™ proof-of-closure system

  • Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
  • Stackable, leak-resistant and graduated unless otherwise noted
  • Sterile containers labeled with a security tab label and sterilized by gamma irradiation to SAL 10-3; certification of sterility available upon request
  • Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used is free from latex and bovine contamination
  • Manufactured under strict process control and lot numbers are fully traceable
  • 95kpa compliant; testing results available upon request
  • Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
  • Available in individually wrapped (Flo-Pac™) formats
  • Closures secure at 10 to 12 in.-lb. closing force
  • Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post-production cap-back-off
  • Standard cap colors are natural, orange and blue

Specifications

Graduated Yes
Material Polypropylene (Clarified)
Sterility Sterile
Capacity (English) 3.0 oz.
Capacity (Metric) 90 mL
Closure Color Natural
Closure Material Polyethylene
Closure Size 53 mm
Label Tamper-evident Label
No. per Case 400/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.