Learn More
PARVA Mouse anti-Canine, Human, Mouse, Clone: OTI2F10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505997

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Members of the parvin family, including PARVA, are actin-binding proteins associated with focal contacts.Specifications
PARVA | |
Monoclonal | |
Unconjugated | |
PARVA | |
CH-ILKBP, MXRA2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
476859, 55742, 57342 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2F10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9EPC1, Q9NVD7 | |
PARVA | |
Full length human recombit protein of human PARVA produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.