Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARVA Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA125074

Catalog No. PIPA125074


Only null left
Add to Cart

Description

Description

Recommended positive controls: ECV304. PA1-25074 is expected to cross react with human (94% conserved) and rat (100% conserved) due to sequence homology. The PA1-25074 immunogen is a synthetic peptide gtlarrkkakevsefqeeg conjugated to KLH, corresponding to amino acids 35-53 of Mouse Parvin alpha. The sequence is identical in human actopaxin and has considerable homology (70%) with beta parvin/affixin, but no homology with gamma parvin.

The protein encoded by this gene is a transcription factor that contains C2H2-type zinc-fingers. It also contains a positive regulatory domain, which has been found in several other zinc-finger transcription factors including those involved in B cell differentiation and tumor suppression. Studies of the mouse counterpart suggest that this protein may be involved in the development of the central nerve system (CNS), as well as in the pathogenesis of neuronal storage disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PARVA
Polyclonal
Unconjugated
PARVA
2010012A22Rik; 5430400F08Rik; Actopaxin; Actp; AI225929; Alpha-parvin; alpha-parvin-like; AU042898; Calponin-like integrin-linked kinase-binding protein; CH-ILKBP; FLJ10793; FLJ12254; I79_014720; Matrix-remodeling-associated protein 2; MNCb-0301; MXRA2; PARVA; Parvin; parvin alpha; parvin, alpha
Rabbit
IgG fraction
RUO
55742, 57342
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
8.3 mg/mL
PBS with 15 mM sodium azide; pH 7.4
Q9EPC1, Q9NVD7
PARVA
Synthetic peptide gtlarrkkakevsefqeeg conjugated to KLH, corresponding to amino acids 35-53 of Mouse Parvin alpha. The sequence is identical in human actopaxin and has considerable homology (70%) with beta parvin/affixin, but no homology with gamma parvin.
50 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.