Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARVA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531182 Supplier Invitrogen™ Supplier No. PA531182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, mouse kidney. Predicted reactivity: Mouse (99%), Rat (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a transcription factor that contains C2H2-type zinc-fingers. It also contains a positive regulatory domain, which has been found in several other zinc-finger transcription factors including those involved in B cell differentiation and tumor suppression. Studies of the mouse counterpart suggest that this protein may be involved in the development of the central nerve system (CNS), as well as in the pathogenesis of neuronal storage disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARVA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PARVA
Gene Accession No. Q9EPC1, Q9NVD7
Gene Alias 2010012A22Rik; 5430400F08Rik; Actopaxin; Actp; AI225929; Alpha-parvin; alpha-parvin-like; AU042898; Calponin-like integrin-linked kinase-binding protein; CH-ILKBP; FLJ10793; FLJ12254; I79_014720; Matrix-remodeling-associated protein 2; MNCb-0301; MXRA2; PARVA; Parvin; parvin alpha; parvin, alpha
Gene Symbols PARVA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Parvin alpha. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55742, 57342
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.