Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARVA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580883
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Actopaxin.
The protein encoded by this gene is a transcription factor that contains C2H2-type zinc-fingers. It also contains a positive regulatory domain, which has been found in several other zinc-finger transcription factors including those involved in B cell differentiation and tumor suppression. Studies of the mouse counterpart suggest that this protein may be involved in the development of the central nerve system (CNS), as well as in the pathogenesis of neuronal storage disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
PARVA | |
Polyclonal | |
Unconjugated | |
PARVA | |
2010012A22Rik; 5430400F08Rik; Actopaxin; Actp; AI225929; Alpha-parvin; alpha-parvin-like; AU042898; Calponin-like integrin-linked kinase-binding protein; CH-ILKBP; FLJ10793; FLJ12254; I79_014720; Matrix-remodeling-associated protein 2; MNCb-0301; MXRA2; PARVA; Parvin; parvin alpha; parvin, alpha | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
55742 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9NVD7 | |
PARVA | |
Recombinant Human Actopaxin protein (Met1-Glu372). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction