Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parvalbumin Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297030G
Description
Parvalbumin Polyclonal specifically detects Parvalbumin in Human samples. It is validated for ELISA.Specifications
Parvalbumin | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 488 | |
D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
Produced in rabbits immunized with purified, recombinant Parvalbumin (P20472; Met1-Ser110). | |
0.1 mL | |
Neuroscience | |
5816 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction