Learn More
Invitrogen™ Parvalbumin Monoclonal Antibody (C12), PerCP

Description
A 1:500 dilution of MA5-45929 was sufficient for detection of Parvalbumin in 15 μg of Rat Brain by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects 12 kDa.
Specifications
Specifications
| Antigen | Parvalbumin |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | C12 |
| Concentration | 1 mg/mL |
| Conjugate | PerCP |
| Formulation | 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 |
| Gene | Pvalb |
| Gene Accession No. | P02625, P20472, P32848 |
| Gene Alias | D22S749; HGNC:9704; MGC116759; PALB1; Parv; parvalbumin; Parvalbumin (calcium binding protein); parvalbumin alpha; PV; Pva; PVALB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.