Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARVB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | PARVB |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1592920
![]() |
Novus Biologicals
NBP15912920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159129
![]() |
Novus Biologicals
NBP159129 |
100 μL |
Each for $499.50
|
|
|||||
Description
PARVB Polyclonal specifically detects PARVB in Human samples. It is validated for Western Blot.Specifications
PARVB | |
Polyclonal | |
Rabbit | |
Cancer | |
affixin, beta-parvin, CGI-56, parvin, beta | |
PARVB | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q96PN1 | |
29780 | |
Synthetic peptides corresponding to PARVB(parvin, beta) The peptide sequence was selected from the N terminal of PARVB. Peptide sequence LQEEGKNAINSPMSPALVDVHPEDTQLEENEERTMIDPTSKEDPKFKELV. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title