Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARVB Antibody (OTI1B7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273248FR
Description
PARVB Monoclonal specifically detects PARVB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PARVB | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PARVB | |
Full length recombinant protein of human PARVB (NP_037459) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1B7 | |
Western Blot, Immunohistochemistry | |
affixin, beta-parvin, CGI-56, parvin, beta | |
Mouse | |
Affinity Purified | |
Cancer | |
29780.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction