Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARVB Mouse anti-Human, Clone: OTI3G1, lyophilized, TrueMAB™
SDP

Catalog No. 501663862
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-3862 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-3862 Supplier OriGene Supplier No. CF505715
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.

Specifications

Antigen PARVB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G1
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PARVB
Gene Accession No. Q9HBI1
Gene Alias CGI-56
Gene Symbols PARVB
Host Species Mouse
Immunogen Full length human recombit protein of human PARVB produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29780
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.