Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARVB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542805 Supplier Invitrogen™ Supplier No. PA542805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LQEEGKNAIN SPMSPALVDV HPEDTQLEEN EERTMIDPTS KEDPKFKELV Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 85%.

Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARVB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Parvb
Gene Accession No. Q9HBI1
Gene Alias Affixin; AI595373; AW742462; beta-parvin; CGI-56; D15Gsk1; fd02e01; hypothetical protein LOC325686; Parvb; parvin beta; parvin, beta; wu:fd02e01; zgc:73117
Gene Symbols Parvb
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PARVB (aa 41-90).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29780
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.