Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARVB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587157
Description
Purity is > 95% by SDS-PAGE.
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
Specifications
PARVB | |
Polyclonal | |
Unconjugated | |
Parvb | |
Affixin; AI595373; AW742462; beta-parvin; CGI-56; D15Gsk1; fd02e01; hypothetical protein LOC325686; Parvb; parvin beta; parvin, beta; wu:fd02e01; zgc:73117 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170736, 29780, 362973 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ES46, Q9HBI1 | |
Parvb | |
Recombinant fusion protein of human PARVB. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction