Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARVB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589039 Supplier Invitrogen™ Supplier No. PA589039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSAPRSPTP RPRRMKKDES FLGKLGGTLA RKRRAREVSD LQEEGKNAIN SPMSPALVDV HPEDTQLEEN EERTMIDPTS KEDPKFKELV KVLLDWINDV; Positive Samples: U-87MG, A-375, HeLa, Mouse heart, Rat heart; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, Z line, cytoskeleton, focal adhesion, lamellipodium, myofibril, sarcomere.

Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARVB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Parvb
Gene Accession No. Q9ES46, Q9HBI1
Gene Alias Affixin; AI595373; AW742462; beta-parvin; CGI-56; D15Gsk1; fd02e01; hypothetical protein LOC325686; Parvb; parvin beta; parvin, beta; wu:fd02e01; zgc:73117
Gene Symbols Parvb
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human PARVB (NP_0374592).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170736, 29780, 362973
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.