Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARVB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106425
Description
Antibody detects endogenous levels of total PARVB.
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
Specifications
| PARVB | |
| Polyclonal | |
| Unconjugated | |
| Parvb | |
| Affixin; AI595373; AW742462; beta-parvin; CGI-56; D15Gsk1; fd02e01; hypothetical protein LOC325686; Parvb; parvin beta; parvin, beta; wu:fd02e01; zgc:73117 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170736, 29780, 362973 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9ES46, Q9HBI1 | |
| Parvb | |
| A synthesized peptide derived from human PARVB(Accession Q9HBI1), corresponding to amino acid residues Y306-L356. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction