Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARVG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565109
Description
Immunogen sequence: RFQPDLSLPTN VQVEVITIES TKSGLKSEKL VEQLTEYSTD KDEPPKDVFD ELFKLAPEKV NAVKEAIVNF VN Highest antigen sequence identity to the following orthologs - mouse 77%, rat 75%.
Probably plays a role in the regulation of cell adhesion and cytoskeleton organization.
Specifications
PARVG | |
Polyclonal | |
Unconjugated | |
Parvg | |
AI413459; Gamma-parvin; gamma-parvin; LOW QUALITY PROTEIN: gamma-parvin; PARVG; parvin gamma; parvin, gamma; zgc:162304 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64098 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBI0 | |
Parvg | |
Recombinant Human PARVG. Recombinant protein control fragment (Product #RP-105868). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction