Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parvin gamma Antibody (8C5.2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308728C
Description
Parvin gamma Monoclonal antibody specifically detects Parvin gamma in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Parvin gamma | |
Monoclonal | |
DyLight 650 | |
gamma-parvin, parvin, gamma | |
GST-tagged recombinant protein corresponding to human Parvin gamma (Uniprot: Q9HBI0) | |
100 μg | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
64098 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Immunoprecipitation | |
8C5.2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction