Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parvin gamma Rabbit anti-Human, Clone: PARVG/6312R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
| Antigen | Parvin gamma |
|---|---|
| Clone | PARVG/6312R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Parvin gamma Monoclonal specifically detects Parvin gamma in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Parvin gamma | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| gamma-parvin, parvin, gamma | |
| GST-tagged recombinant protein corresponding to human Parvin gamma | |
| Primary | |
| Store at 4°C. |
| PARVG/6312R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 64098 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title