Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parvovirus B19 Monoclonal Antibody (5191)
GREENER_CHOICE

Catalog No. PIMA533288
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533288 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533288 Supplier Invitrogen™ Supplier No. MA533288
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Negative in uninfected human cells.

Parvoviruses cause infections in a wide variety of birds and mammals. Parvovirus, commonly called parvo, is a genus of the Parvoviridae family linear, non-segmented single stranded DNA viruses with an average genome size of 5 kbp. Parvoviruses are some of the smallest viruses found in nature. Parvovirus B19 infection is common among children and adults and is often asymptomatic. Parvovirus B19 infection can result in serious and occasionally fatal hematologic diseases in susceptible patients, and is the cause of fifth disease, a highly contagious infection of childhood. Parvovirus B19 can also infect the fetus causing severe anemia, congestive heart failure, and death in utero or at birth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parvovirus B19
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 5191
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.