Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvovirus Mouse anti-Dog, Clone: 1C10, Abnova™

Catalog No. 89933719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-719 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-719 Supplier Abnova Corporation Supplier No. MAB12434

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against dog Parvovirus.

Specifications

Antigen Parvovirus
Applications ELISA
Classification Monoclonal
Clone 1C10
Conjugate Unconjugated
Formulation Lyophilized from PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Canine Parvovirus
Purification Method Antigen Affinity Purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at −20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.