Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PASD1 Antibody (2ALCC136), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250235V
Description
PASD1 Monoclonal specifically detects PASD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
PASD1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PASD1 | |
GST-PASD1 recombinant protein containing 195-474 aas of PASD1. Region is common to PASD1a and PASD1b proteins. | |
0.1 mL | |
Epigenetics | |
139135 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
2ALCC136 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CT63Cancer/testis antigen 63, OXTES1, OX-TES-1, PAS domain containing 1, PAS domain-containing protein 1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
PASD1-1 recognizes the recombinant protein (aa 195-474) present in both PASD1a and PASD1b proteins. | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction