Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PASD1 Antibody (2ALCC136), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309733 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309733 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309733 Supplier Novus Biologicals Supplier No. NBP250235MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PASD1 Monoclonal antibody specifically detects PASD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PASD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2ALCC136
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CT63Cancer/testis antigen 63, OXTES1, OX-TES-1, PAS domain containing 1, PAS domain-containing protein 1
Host Species Mouse
Immunogen GST-PASD1 recombinant protein containing 195-474 aas of PASD1. Region is common to PASD1a and PASD1b proteins.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 139135
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.