Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PASD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531463 Supplier Invitrogen™ Supplier No. PA531463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3, SK-N-SH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is thought to function as a transcription factor. The protein is a cancer-associated antigen that can stimulate autologous T-cell responses, and it is therefore considered to be a potential immunotherapeutic target for the treatment of various hematopoietic malignancies, including diffuse large B-cell lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PASD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene PASD1
Gene Accession No. Q8IV76
Gene Alias Cancer/testis antigen 63; circadian clock protein PASD1; CT63; Gm1141; OXTES1; OX-TES-1; PAS domain containing 1; PAS domain-containing protein 1; PASD1; predicted gene 1141
Gene Symbols PASD1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PASD1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.