Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PASD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552800 Supplier Invitrogen™ Supplier No. PA552800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEKDEVYQKI ILKFPLLNSE THIEFCCHLK RGNVEHGDSS AYENVKFIVN VRDICNEFPV VFSGLFSSHL CADFAACVPQ EDRLYLVGNV CILRTQLLQQ LYTSKAVSDE AVLTQDSDEE PFVGELSSSQ GQRGHTSMKA VYVEPAAA Highest antigen sequence identity to the following orthologs: Mouse - 19%, Rat - 19%.

This gene encodes a protein that is thought to function as a transcription factor. The protein is a cancer-associated antigen that can stimulate autologous T-cell responses, and it is therefore considered to be a potential immunotherapeutic target for the treatment of various hematopoietic malignancies, including diffuse large B-cell lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PASD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PASD1
Gene Accession No. Q8IV76
Gene Alias Cancer/testis antigen 63; circadian clock protein PASD1; CT63; Gm1141; OXTES1; OX-TES-1; PAS domain containing 1; PAS domain-containing protein 1; PASD1; predicted gene 1141
Gene Symbols PASD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PASD1. Recombinant protein control fragment (Product #RP-92229).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.