Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PASD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113119
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113119 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113119 Supplier Invitrogen™ Supplier No. PA5113119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PASD1 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a protein that is thought to function as a transcription factor. The protein is a cancer-associated antigen that can stimulate autologous T-cell responses, and it is therefore considered to be a potential immunotherapeutic target for the treatment of various hematopoietic malignancies, including diffuse large B-cell lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PASD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PASD1
Gene Accession No. Q8IV76
Gene Alias Cancer/testis antigen 63; circadian clock protein PASD1; CT63; Gm1141; OXTES1; OX-TES-1; PAS domain containing 1; PAS domain-containing protein 1; PASD1; predicted gene 1141
Gene Symbols PASD1
Host Species Rabbit
Immunogen Recombinant Human Circadian clock protein PASD1 protein (301-600AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139135
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.