Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PASK Antibody (OTI8G9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273249H
Description
PASK Monoclonal specifically detects PASK in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PASK | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PASK | |
Full length recombinant protein of human PASK (NP_055963) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI8G9 | |
Western Blot, Immunohistochemistry | |
DKFZp434O051, EC 2.7.11.1, hPASK, KIAA0135DKFZp686P2031, PAS domain containing serine/threonine kinase, PAS-kinase, PASKINPAS domain-containing serine/threonine-protein kinase, PAS-serine/threonine kinase, STK37 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
23178.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction