Learn More
Promega PASK Kinase Enzyme System, 10microg
Catalog No. PRV4240 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4240 10 μg
1 options
Catalog No. PRV4240 Supplier Promega Supplier No. V4240
Recombinant human PASK (981-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PASK or PAS domain containing serine/threonine kinase regulates the function of many intracellular signaling pathways involved in stress.
  • Easily Screen and Profile PASK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C