Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PASK Antibody (OTI8G9), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273249AF594
Description
PASK Monoclonal specifically detects PASK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| PASK | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PASK | |
| Full length recombinant protein of human PASK (NP_055963) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI8G9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp434O051, EC 2.7.11.1, hPASK, KIAA0135DKFZp686P2031, PAS domain containing serine/threonine kinase, PAS-kinase, PASKINPAS domain-containing serine/threonine-protein kinase, PAS-serine/threonine kinase, STK37 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 23178.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction