Learn More
PASK Mouse anti-Human, Clone: OTI11D8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500878

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PASKIN (PAS-Kinase) is thought to regulate protein synthesis in cells. It is a conserved gene product found in yeast as well as mammals. PASKIN contains two domains (PAS A and PAS B) as well as a serine/threonine kinase domain related to AMP kinases. PASKIN activity leads to decreased carbohydrate storage, as well as increased protein synthesis. PASKIN-dependent phosphorylation inhibits the activity of mammalian glycogen synthase.Specifications
PASK | |
Monoclonal | |
Unconjugated | |
PASK | |
PASKIN, STK37 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23178 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI11D8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96RG2 | |
PASK | |
Full length human recombit protein of human PASK produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.