Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PASK Mouse anti-Human, Clone: OTI8G9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500826

Description
PASKIN (PAS-Kinase) is thought to regulate protein synthesis in cells. It is a conserved gene product found in yeast as well as mammals. PASKIN contains two domains (PAS A and PAS B) as well as a serine/threonine kinase domain related to AMP kinases. PASKIN activity leads to decreased carbohydrate storage, as well as increased protein synthesis. PASKIN-dependent phosphorylation inhibits the activity of mammalian glycogen synthase.Specifications
PASK | |
Monoclonal | |
0.93 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96RG2 | |
PASK | |
Full length human recombit protein of human PASK produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI8G9 | |
Unconjugated | |
PASK | |
PASKIN, STK37 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23178 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction