Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PASK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582744 Supplier Invitrogen™ Supplier No. PA582744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQEVDVNSFS WNLKELFFSD QTDQTSSNCS CATSELRETP SSLAVGSDPD VGSLQEQGSC VLDDRELLLL TGTCVDLGQG RRFRESCVGH DPTEPLEVCL VSSEH Highest antigen sequence indentity to the following orthologs: Mouse - 64%, Rat - 60%.

PASK is an evolutionarily conserved protein present in yeast, flies, and mammals. PAS domains regulate the function of many intracellular signaling pathways in response to both extrinsic and intrinsic stimuli. It is a conserved gene product found in yeast as well as mammals. PASKIN contains two domains (PAS A and PAS B) as well as a serine/threonine kinase domain related to AMP kinases. PASKIN activity leads to decreased carbohydrate storage, as well as increased protein synthesis. PASKIN-dependent phosphorylation inhibits the activity of mammalian glycogen synthase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PASK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PASK
Gene Accession No. Q96RG2
Gene Alias Kiaa0135; mKIAA0135; OTTMUSP00000024404; PAS domain containing serine/threonine kinase; PAS domain-containing serine/threonine-protein kinase; PASK; PASKIN; PAS-kinase; per-arnt-sim (PAS) domain kinase; STK37
Gene Symbols PASK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PASK. Recombinant protein control fragment (Product #RP-91413).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.