Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PASTICCINO 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114340 Supplier Invitrogen™ Supplier No. PA5114340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody specifically recognizes PASTICCINO 2 in Arabidopsis thaliana.

Catalyzes the third of the four reactions of the long-chain fatty acids elongation cycle. This endoplasmic reticulum-bound enzymatic process, allows the addition of two carbons to the chain of long- and very long-chain fatty acids/VLCFAs per cycle. This enzyme catalyzes the dehydration of the 3-hydroxyacyl-CoA intermediate into trans-2,3-enoyl-CoA, within each cycle of fatty acid elongation. Thereby, it participates in the production of VLCFAs of different chain lengths that are involved in multiple biological processes as precursors of membrane lipids and lipid mediators. May be an anti-phosphatase that prevents CDKA-1 dephosphorylation and activation. Involved in the hormonal control of cell division and differentiation. Required for proliferation control of meristematic and non-meristematic cells. Negative regulator of the cell cycle. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PASTICCINO 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene PAS2
Gene Accession No. Q8VZB2
Gene Alias 3-hydroxyacyl-CoA dehydratase PASTICCINO 2; At5g10480; AtPAS2; F12B17.170; F12B17_170; PAS2; PASTICCINO 2; PASTICCINO2; PEP; PEPINO; PEPINO protein; Protein PEPINO; Protein tyrosine phosphatase-like protein; Protein-tyrosine phosphatase-like, PTPLA; putative protein phosphatase; Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase PASTICCINO 2
Gene Symbols PAS2
Host Species Rabbit
Immunogen Synthetic peptide within thaliana PAS2 aa 172-221.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 830912
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.