Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531005
Description
Recommended positive controls: Jurkat, Raji. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (96%), Xenopus laevis (86%), Chicken (98%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene interacts with microtubules and is functionally associated with beta-amyloid precursor protein transport and/or processing. The beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. This gene has been found to be highly expressed in breast cancer. Multiple polyadenylation sites have been found for this gene.
Specifications
PAT1 | |
Polyclonal | |
Unconjugated | |
APPBP2 | |
1300003O07Rik; AI465480; amyloid beta precursor protein (cytoplasmic tail) binding protein 2; amyloid beta precursor protein binding protein 2; amyloid beta precursor protein-binding protein 2; Amyloid protein-binding protein 2; Appbp2; APP-BP2; appbp2 {ECO:0000312; EMBL:ABQ00241.1}; HS.84084; KIAA0228; PAT1; Protein interacting with APP tail 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10513 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.44 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q92624 | |
APPBP2 | |
Recombinant fragment corresponding to a region within amino acids 327 and 575 of Human APPBP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction