Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572784
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572784 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572784 Supplier Invitrogen™ Supplier No. PA572784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by this gene interacts with microtubules and is functionally associated with beta-amyloid precursor protein transport and/or processing. The beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. This gene has been found to be highly expressed in breast cancer. Multiple polyadenylation sites have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAT1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene APPBP2
Gene Accession No. A5HK05, Q92624, Q9DAX9
Gene Alias 1300003O07Rik; AI465480; amyloid beta precursor protein (cytoplasmic tail) binding protein 2; amyloid beta precursor protein binding protein 2; amyloid beta precursor protein-binding protein 2; Amyloid protein-binding protein 2; Appbp2; APP-BP2; appbp2 {ECO:0000312; EMBL:ABQ00241.1}; HS.84084; KIAA0228; PAT1; Protein interacting with APP tail 1
Gene Symbols APPBP2
Host Species Rabbit
Immunogen APPBP2 antibody for an 18 amino acid peptide near the carboxy terminus of human APPBP2. The immunogen is within the last 50 amino acids of APPBP2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10513, 303396, 66884
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.