Learn More
Invitrogen™ PAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590700
Description
Immunogen sequence: LEEIAIDCHN KETEQRLLQE AHDLHLSSLQ LAKKAFGEFN VQTAKHYGNL GRLYQSMRKF KEAEEMHIKA IQIKEQLLGQ EDYEVALSVG HLASLYNYDM NQYENAEKLY LRSIAIGKKL FGEGYSGLEY DYRGLIKLYN SIGNYEKVFE YHNVLSNWNR LRDRQYSVTD ALEDVSTSPQ STEEVVQSFL ISQNVEGPSC; Positive Samples: A-431, HeLa, Jurkat, Raji, Mouse kidney, Mouse brain, Rat kidney, Rat brain; Cellular Location: Cytoplasm, Membrane, Nucleus, Peripheral membrane protein, cytoskeleton.
The protein encoded by this gene interacts with microtubules and is functionally associated with beta-amyloid precursor protein transport and/or processing. The beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. This gene has been found to be highly expressed in breast cancer. Multiple polyadenylation sites have been found for this gene.
Specifications
PAT1 | |
Polyclonal | |
Unconjugated | |
APPBP2 | |
1300003O07Rik; AI465480; amyloid beta precursor protein (cytoplasmic tail) binding protein 2; amyloid beta precursor protein binding protein 2; amyloid beta precursor protein-binding protein 2; Amyloid protein-binding protein 2; Appbp2; APP-BP2; appbp2 {ECO:0000312; EMBL:ABQ00241.1}; HS.84084; KIAA0228; PAT1; Protein interacting with APP tail 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10513, 303396, 66884 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.7 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
A5HK05, Q92624, Q9DAX9 | |
APPBP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 386-585 of human APPBP2 (NP_0063712). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.