Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PATE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585005 Supplier Invitrogen™ Supplier No. PA585005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSGSLSMRND AVNEIVAVKN NFPVIEIVRC RMCHLQFPGE KCSRGRGICT ATTEE Highest antigen sequence indentity to the following orthologs: Rat - 56%, Mouse - 58%.

PATE1 is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PATE1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PATE1
Gene Accession No. Q8WXA2
Gene Alias expressed in prostate and testis; PATE; PATE1; prostate and testis expressed 1; prostate and testis expressed protein 1
Gene Symbols PATE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PATE1. Recombinant protein control fragment (Product #RP-108666).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160065
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.