Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PATE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596096 Supplier Invitrogen™ Supplier No. PA596096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNKHFLFLFL LYCLIVAVTS LQCITCHLRT RTDRCRRGFG VCTAQKGEAC MLLRIYQRNT LQISYMVCQK FCRDMTFDLR NRTYVHTCCN YNYCNFKL; Positive Samples: HeLa.

PATE3 (prostate and testis expressed protein 3), also known as PATE-DJ or HEL-127, is a 98 amino acid protein that contains one UPAR/Ly6 domain and belongs to the PATE family. PATE3 is a secreted protein that is expressed in prostate and testis. The gene that encodes PATE3 consists of around 3,490 bases and maps to human chromosome 11p15.5. Chromosome 11, which comprises approximately 4% of the human genome, is considered a gene and disease association-dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. The blood disorders Sickle cell anemia and thalassemia are caused by HBB gene mutations, while Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11-encoded genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PATE3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PATE3
Gene Accession No. B3GLJ2
Gene Alias Acrosomal vesicle protein HEL-127; epididymis-specific ESC112; Gm17365; HEL-127; Pate3; PATE-DJ; PATE-like protein DJ; predicted gene, 17365; prostate and testis expressed 3; prostate and testis expressed DJ; prostate and testis expressed protein 3; secreted TFP/Ly-6/uPAR protein PATE-DJ
Gene Symbols PATE3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-98 of human PATE3 (NP_0011233553).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100169851
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.