Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PATE4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561056 Supplier Invitrogen™ Supplier No. PA561056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLKCNTCIYT EGWKCMAGRG TCIAKENELC STTAYFRGDK HMYSTHMCKY KCREEESSKR GLLRVTLCGD RNFCNVF Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 35%.

PATE4 may modulate the function of nicotinic acetylcholine receptors and may enhance sperm motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PATE4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PATE4
Gene Accession No. P0C8F1
Gene Alias PATE4; PATE-B; PATE-like protein B; prostate and testis expressed 4; prostate and testis expressed protein 4; prostate and testis expression B
Gene Symbols PATE4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PATE4. Recombinant protein control fragment (Product #RP-100570).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.