Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Path-ID™ Multiplex One-Step RT-PCR Kit

Catalog No. p-7181610
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
44-421-36 500 Reactions
44-421-35 100 Reactions
44-421-37 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 44-421-36 Supplier Applied Biosystems™ Supplier No. 4442136
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Path-ID™ Multiplex One-Step RT-PCR Kit is designed for multiplex, quantitative, reverse transcription PCR (qRT-PCR).

The Path-ID™ Multiplex One-Step RT-PCR Kit is optimized for the amplification of up to four targets simultaneously using your RNA samples and TaqMan™ primer-probe sets. Reactions are run using a single-tube, one-step procedure to reverse-transcribe the RNA and amplify your targets. The kit contains a Multiplex Enzyme Mix with Arrayscript™ reverse transcriptase and AmpliTaq Gold™ DNA Polymerase, and a buffer mix.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Includes 500 reactions with the following components: Multiplex RT-PCR Buffer, Multiplex Enzyme Mix, and Nuclease-free water.
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance Low
PCR Method 1-step RT-qPCR, Multiplex qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 Fast System, 7500 System
Passive Reference Dye ROX (Pre-mixed)
Product Line Path-ID
Product Type Multiplex One-Step RT-PCR Kit
Quantity 500 reactions
Reverse Transcriptase ArrayScript™ UP
Sufficient For 500 Reactions
For Use With (Application) Pathogen Detection, Virus Detection
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Sample Type RNA
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.