Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Path-ID™ qPCR Master Mix

Catalog No. 4388644
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
43-886-44 500 Reactions
43-886-43 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-886-44 Supplier Applied Biosystems™ Supplier No. 4388644
  • PRODUCT PROMOTION
    $1,012.00
    $755.00 / Each of 1

    Save $257.00

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection of DNA targets.

Ambion™ Path-ID™ qPCR Master Mix is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:

  • AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
  • Optimized buffer and dNTP concentration, for reliable PCR
  • Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Primer-probe
Form Liquid
Format Tube
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line Ambion, Path-ID
Purity or Quality Grade UP (Ultra Pure)
Quantity 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Animal Health, Pathogen Detection
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Veterinary Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.