Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polymedco PATHFAST Immunoassay Reagent Cardiac Troponin I
Shop All Polymedco Products

Click to view available options
:
Cardiac Troponin I Reagent
Description
- Includes 60 tests
Specifications
Specifications
CE Marker | CE-IVD |
Clia Complexity | Moderate (Serum, Plasma or Whole Blood), Waived (Other) |
Disposable | Single-use |
No. of Reactions | 60 |
Form | Ready-to-Use (RTU) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction