Learn More
StatLab PathFlow Connected Lid Cassettes Slotted
SDP
Catalog No. 21033296
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Pore Style:
Slotted Loose
Slotted Taped
Catalog No. 21-033-296
Supplier: StatLab CAS505YLT
Only null left
Add to Cart
Add to Cart
One Piece design with rear hinge and front tab closure
  • The 35 degree print face is designed to be compatible for printing on the Leica IP C Inkjet Cassette Printer, Primera Signature cassette printer and the Tissue-Tek SmartWrite Cassette Printer
  • These cassettes are a suitable replacement for the Unisette (M505) and Signature cassettes(M405)
  • Loose, 1500/case
  • Taped, 1000/case
Specifications
Pore Style Slotted Taped
Color Yellow
Product Type Embedding Cassettes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.