Learn More
StatLab PathFlow PiSmart Laser Attached Lid Front Hinge Slotted Embedding Cassettes
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
Description
- Slotted embedding cassettes
- Front hinge with lid attached
- Made from acetal polymer
- Specifically validated for the Pi Smart Laser Cassette Printer
- Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
- Threaded
- 2000 per case
Specifications
Specifications
Quantity | 2000 Cassettes/Cs. |
Color | Blue |
Product Type | Slotted Embedding Cassettes |
Made in USA